Anti-STX18 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00053407-D01P
103344-586EA 844.62 USD
103344-586
Anti-STX18 Rabbit Polyclonal Antibody
Antibodies
The STX18 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STX18. This antibody reacts with human. The STX18 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: STX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR