Anti-STX11 Mouse Monoclonal Antibody [clone: 4F9]

Supplier: Novus Biologicals
H00008676-M01
103335-422EA 798.46 USD
103335-422
Anti-STX11 Mouse Monoclonal Antibody [clone: 4F9]
Antibodies
The STX11 Antibody (4F9) from Novus Biologicals is a mouse monoclonal antibody to STX11. This antibody reacts with human. The STX11 Antibody (4F9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: STX11
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR