Anti-STT3A Mouse Monoclonal Antibody [clone: 4D4]

Supplier: Novus Biologicals
H00003703-M02
103329-290EA 798.46 USD
103329-290
Anti-STT3A Mouse Monoclonal Antibody [clone: 4D4]
Antibodies
The STT3A Antibody (4D4) from Novus Biologicals is a mouse monoclonal antibody to STT3A. This antibody reacts with human. The STT3A Antibody (4D4) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STT3A
Clonality: Monoclonal
Clone: 4D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR