Anti-STRBP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20522
102711-914EA 635.51 USD
102711-914
Anti-STRBP Rabbit Polyclonal Antibody
Antibodies
The STRBP Antibody from Novus Biologicals is a rabbit polyclonal antibody to STRBP. This antibody reacts with human. The STRBP Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: STRBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR