Anti-STN2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90658
103277-938EA 777.05 USD
103277-938
Anti-STN2 Rabbit Polyclonal Antibody
Antibodies
The stonin-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to stonin-2. This antibody reacts with human. The stonin-2 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR