Anti-STK4 Mouse Monoclonal Antibody [clone: 4F4]

Supplier: Novus Biologicals
H00006789-M02
103310-942EA 798.46 USD
103310-942
Anti-STK4 Mouse Monoclonal Antibody [clone: 4F4]
Antibodies
The STK4 Antibody (4F4) from Novus Biologicals is a mouse monoclonal antibody to STK4. This antibody reacts with human, mouse, rat. The STK4 Antibody (4F4) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Proximity Ligation Assay.

Type: Primary
Antigen: STK4
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR