Anti-STK38L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20518
102711-906EA 635.51 USD
102711-906
Anti-STK38L Rabbit Polyclonal Antibody
Antibodies
The STK38L Antibody from Novus Biologicals is a rabbit polyclonal antibody to STK38L. This antibody reacts with human. The STK38L Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: STK38L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR