Anti-STK38L Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00023012-B01P
103340-882EA 399 USD
103340-882
Anti-STK38L Mouse Polyclonal Antibody
Antibodies
The STK38L Antibody from Novus Biologicals is a mouse polyclonal antibody to STK38L. This antibody reacts with human. The STK38L Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: STK38L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR