Anti-STK38L Mouse Monoclonal Antibody [clone: 4E5]

Supplier: Novus Biologicals
H00023012-M01
103340-884EA 798.46 USD
103340-884
Anti-STK38L Mouse Monoclonal Antibody [clone: 4E5]
Antibodies
The STK38L Antibody (4E5) from Novus Biologicals is a mouse monoclonal antibody to STK38L. This antibody reacts with human. The STK38L Antibody (4E5) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STK38L
Clonality: Monoclonal
Clone: 400000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR