Anti-STK36 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83411
103267-702EA 734.58 USD
103267-702
Anti-STK36 Rabbit Polyclonal Antibody
Antibodies
The STK36 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STK36. This antibody reacts with human. The STK36 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STK36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR