Anti-STK32C Mouse Monoclonal Antibody [clone: 4D12]

Supplier: Novus Biologicals
H00282974-M03
103348-040EA 798.46 USD
103348-040
Anti-STK32C Mouse Monoclonal Antibody [clone: 4D12]
Antibodies
The STK32C Antibody (4D12) from Novus Biologicals is a mouse monoclonal antibody to STK32C. This antibody reacts with human, mouse. The STK32C Antibody (4D12) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: STK32C
Clonality: Monoclonal
Clone: 4D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR