Anti-STK32A Mouse Monoclonal Antibody [clone: 1G6]

Supplier: Novus Biologicals
H00202374-M03
103347-736EA 399 USD
103347-736
Anti-STK32A Mouse Monoclonal Antibody [clone: 1G6]
Antibodies
The STK32A Antibody (1G6) from Novus Biologicals is a mouse monoclonal antibody to STK32A. This antibody reacts with human. The STK32A Antibody (1G6) has been validated for the following applications: ELISA.

Type: Primary
Antigen: STK32A
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR