Anti-STK19 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00008859-D01P
103335-806EA 844.62 USD
103335-806
Anti-STK19 Rabbit Polyclonal Antibody
Antibodies
The STK19 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STK19. This antibody reacts with human. The STK19 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: STK19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR