Anti-STI1 Mouse Monoclonal Antibody [clone: 20]

Supplier: Novus Biologicals
H00010963-M33
103339-892EA 798.46 USD
103339-892
Anti-STI1 Mouse Monoclonal Antibody [clone: 20]
Antibodies
The STI1 Antibody (2E1) from Novus Biologicals is a mouse monoclonal antibody to STI1. This antibody reacts with human. The STI1 Antibody (2E1) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STI1
Clonality: Monoclonal
Clone: 20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR