Anti-STAU2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-89380
103276-574EA 734.58 USD
103276-574
Anti-STAU2 Rabbit Polyclonal Antibody
Antibodies
The STAU2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STAU2. This antibody reacts with human, mouse, rat. The STAU2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STAU2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR