Anti-STAU2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
30890002-0.1MG
103317-206EA 349 USD
103317-206
Anti-STAU2 Rabbit Polyclonal Antibody
Antibodies
The STAU2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STAU2. This antibody reacts with human. The STAU2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STAU2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR