Anti-STAU2 Mouse Monoclonal Antibody [clone: 1B9]

Supplier: Novus Biologicals
H00027067-M04
103342-530EA 798.46 USD
103342-530
Anti-STAU2 Mouse Monoclonal Antibody [clone: 1B9]
Antibodies
The STAU2 Antibody (1B9) from Novus Biologicals is a mouse monoclonal antibody to STAU2. This antibody reacts with human. The STAU2 Antibody (1B9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: STAU2
Clonality: Monoclonal
Clone: 1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR