Anti-STAT6 Mouse Monoclonal Antibody [clone: 4G7]

H00006778-M09
103310-890EA 583.08 USD
103310-890
Anti-STAT6 Mouse Monoclonal Antibody [clone: 4G7]
The STAT6 Antibody (4G7) from Novus Biologicals is a mouse monoclonal antibody to STAT6. This antibody reacts with human. The STAT6 Antibody (4G7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: STAT6
Clonality: Monoclonal
Clone: 4G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR