Anti-STAT5b Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
26980002-0.1MG
103316-892EA 349 USD
103316-892
Anti-STAT5b Rabbit Polyclonal Antibody
Antibodies
The STAT5b Antibody from Novus Biologicals is a rabbit polyclonal antibody to STAT5b. This antibody reacts with human. The STAT5b Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STAT5b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR