Anti-STAT5b Mouse Monoclonal Antibody [clone: 1C2]

H00006777-M04
103310-882EA 583.08 USD
103310-882
Anti-STAT5b Mouse Monoclonal Antibody [clone: 1C2]
The STAT5b Antibody (1C2) from Novus Biologicals is a mouse monoclonal antibody to STAT5b. This antibody reacts with human. The STAT5b Antibody (1C2) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.

Type: Primary
Antigen: STAT5b
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR