Anti-STAT5b Mouse Monoclonal Antibody [clone: 5D6]

H00006777-M02
103310-878EA 583.08 USD
103310-878
Anti-STAT5b Mouse Monoclonal Antibody [clone: 5D6]
The STAT5b Antibody (5D6) from Novus Biologicals is a mouse monoclonal antibody to STAT5b. This antibody reacts with human. The STAT5b Antibody (5D6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STAT5b
Clonality: Monoclonal
Clone: 5D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR