Anti-STAT5a Mouse Monoclonal Antibody [clone: 1B12]

Supplier: Novus Biologicals
H00006776-M02
103310-872EA 767.69 USD
103310-872
Anti-STAT5a Mouse Monoclonal Antibody [clone: 1B12]
Antibodies
The STAT5a Antibody (1B12) from Novus Biologicals is a mouse monoclonal antibody to STAT5a. This antibody reacts with human. The STAT5a Antibody (1B12) has been validated for the following applications: Western Blot, ELISA, Proximity Ligation Assay.

Type: Primary
Antigen: STAT5a
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR