Anti-STAT1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56314SS
103358-798EA 208 USD
103358-798
Anti-STAT1 Rabbit Polyclonal Antibody
Antibodies
The STAT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STAT1. This antibody reacts with human, mouse, porcine, canine, primate. The STAT1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Porcine, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR