Anti-STAT1 Rabbit Polyclonal Antibody

H00006772-D01P
103310-850EA 627.69 USD
103310-850
Anti-STAT1 Rabbit Polyclonal Antibody
The STAT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STAT1. This antibody reacts with human, mouse. The STAT1 Antibody has been validated for the following applications: Western Blot, Proximity Ligation Assay.

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR