Anti-STAT1 Mouse Monoclonal Antibody [clone: 1A8]

H00006772-M01
103310-852EA 553.85 USD
103310-852
Anti-STAT1 Mouse Monoclonal Antibody [clone: 1A8]
The STAT1 Antibody (1A8) from Novus Biologicals is a mouse monoclonal antibody to STAT1. This antibody reacts with human. The STAT1 Antibody (1A8) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR