Anti-STARD8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-32567
103282-426EA 734.58 USD
103282-426
Anti-STARD8 Rabbit Polyclonal Antibody
Antibodies
The STARD8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STARD8. This antibody reacts with human. The STARD8 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STARD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR