Anti-STARD3 Mouse Monoclonal Antibody [clone: 1D1]

Supplier: Novus Biologicals
H00010948-M01
103339-864EA 798.46 USD
103339-864
Anti-STARD3 Mouse Monoclonal Antibody [clone: 1D1]
Antibodies
The STARD3 Antibody (1D1) from Novus Biologicals is a mouse monoclonal antibody to STARD3. This antibody reacts with human. The STARD3 Antibody (1D1) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: STARD3
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR