Anti-STARD10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-84508
103260-428EA 734.58 USD
103260-428
Anti-STARD10 Rabbit Polyclonal Antibody
Antibodies
The STARD10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to STARD10. This antibody reacts with human. The STARD10 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: STARD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR