Anti-StAR Mouse Monoclonal Antibody [clone: 2B5]

Supplier: Novus Biologicals
H00006770-M10
103310-846EA 798.46 USD
103310-846
Anti-StAR Mouse Monoclonal Antibody [clone: 2B5]
Antibodies
The StAR Antibody (2B5) from Novus Biologicals is a mouse monoclonal antibody to StAR. This antibody reacts with human. The StAR Antibody (2B5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: StAR
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR