Anti-STAM1 Mouse Monoclonal Antibody (Biotin) [clone: 29C678]

Supplier: Novus Biologicals
NB100-56533B
103359-850EA 614.15 USD
103359-850
Anti-STAM1 Mouse Monoclonal Antibody (Biotin) [clone: 29C678]
Antibodies
The STAM-1 Antibody (29C678) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to STAM-1. This antibody reacts with human. The STAM-1 Antibody (29C678) [Biotin] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: STAM1
Clonality: Monoclonal
Clone: 29C678
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR