Anti-STAG2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-60946
103409-188EA 564.74 USD
103409-188
Anti-STAG2 Goat Polyclonal Antibody
Antibodies
The STAG2 Antibody from Novus Biologicals is a goat polyclonal antibody to STAG2. This antibody reacts with human. The STAG2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Peptide ELISA.

Type: Primary
Antigen: STAG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: REPKRLRPEDS
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR