Anti-ST8SIA4 Mouse Monoclonal Antibody [clone: 1H5]

Supplier: Novus Biologicals
H00007903-M03
103334-508EA 798.46 USD
103334-508
Anti-ST8SIA4 Mouse Monoclonal Antibody [clone: 1H5]
Antibodies
The ST8 alpha-2,8-Sialyltransferase 4 / ST8SIA4 Antibody (1H5) from Novus Biologicals is a mouse monoclonal antibody to ST8 alpha-2,8-Sialyltransferase 4 / ST8SIA4. This antibody reacts with human. The ST8 alpha-2,8-Sialyltransferase 4 / ST8SIA4 Antibody (1H5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: ST8SIA4
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR