Anti-ST6GALNAC2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00010610-B01P
103339-258EA 399 USD
103339-258
Anti-ST6GALNAC2 Mouse Polyclonal Antibody
Antibodies
The ST6 Sialyltransferase 2 / ST6GALNAC2 Antibody from Novus Biologicals is a mouse polyclonal antibody to ST6 Sialyltransferase 2 / ST6GALNAC2. This antibody reacts with human. The ST6 Sialyltransferase 2 / ST6GALNAC2 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: ST6GALNAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR