Anti-ST6GALNAC1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20493
102711-856EA 635.51 USD
102711-856
Anti-ST6GALNAC1 Rabbit Polyclonal Antibody
Antibodies
The ST6GALNAC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ST6GALNAC1. This antibody reacts with human. The ST6GALNAC1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ST6GALNAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR