Anti-ST3GAL4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13390
103285-764EA 734.58 USD
103285-764
Anti-ST3GAL4 Rabbit Polyclonal Antibody
Antibodies
The ST3GAL4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ST3GAL4. This antibody reacts with human. The ST3GAL4 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ST3GAL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR