Anti-ST3GAL4 Mouse Monoclonal Antibody [clone: 1F4]

Supplier: Novus Biologicals
H00006484-M01
103310-166EA 798.46 USD
103310-166
Anti-ST3GAL4 Mouse Monoclonal Antibody [clone: 1F4]
Antibodies
The ST3GAL4 Antibody (1F4) from Novus Biologicals is a mouse monoclonal antibody to ST3GAL4. This antibody reacts with human. The ST3GAL4 Antibody (1F4) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: ST3GAL4
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR