Anti-ST3GAL2 Mouse Monoclonal Antibody [clone: 1E12.]

Supplier: Novus Biologicals
H00006483-M01
103310-164EA 798.46 USD
103310-164
Anti-ST3GAL2 Mouse Monoclonal Antibody [clone: 1E12.]
Antibodies
The ST3 beta-Gal alpha-2,3-Sialyltransferase 2 / ST3GAL2 Antibody (1E12.) from Novus Biologicals is a mouse monoclonal antibody to ST3 beta-Gal alpha-2,3-Sialyltransferase 2 / ST3GAL2. This antibody reacts with human. The ST3 beta-Gal alpha-2,3-Sialyltransferase 2 / ST3GAL2 Antibody (1E12.) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: ST3GAL2
Clonality: Monoclonal
Clone: 1E12.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR