Anti-ST2/IL-1 R4 Sheep Polyclonal Antibody

NBP2-33319
103282-868EA 493.85 USD
103282-868
Anti-ST2/IL-1 R4 Sheep Polyclonal Antibody
The ST2 / IL-1 R4 Antibody from Novus Biologicals is a sheep polyclonal antibody to ST2 / IL-1 R4. This antibody reacts with human. The ST2 / IL-1 R4 Antibody has been validated for the following applications: Western Blot, Blocking / Neutralizing.

Type: Primary
Antigen: ST2/IL-1 R4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR