Anti-SSX2IP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-85956
103272-334EA 734.58 USD
103272-334
Anti-SSX2IP Rabbit Polyclonal Antibody
Antibodies
The SSX2IP Antibody from Novus Biologicals is a rabbit polyclonal antibody to SSX2IP. This antibody reacts with human. The SSX2IP Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SSX2IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR