Anti-SSX2 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: Novus Biologicals
H00006757-M01
103310-820EA 798.46 USD
103310-820
Anti-SSX2 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
The SSX2 Antibody (1A4) from Novus Biologicals is a mouse monoclonal antibody to SSX2. This antibody reacts with human. The SSX2 Antibody (1A4) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SSX2
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR