Anti-SSPN Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00008082-B02P
103334-654EA 798.46 USD
103334-654
Anti-SSPN Mouse Polyclonal Antibody
Antibodies
The SSPN Antibody from Novus Biologicals is a mouse polyclonal antibody to SSPN. This antibody reacts with human. The SSPN Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SSPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR