Anti-SSH1 Mouse Monoclonal Antibody [clone: 1D2]

Supplier: Novus Biologicals
H00054434-M02
103344-732EA 798.46 USD
103344-732
Anti-SSH1 Mouse Monoclonal Antibody [clone: 1D2]
Antibodies
The SSH1 Antibody (1D2) from Novus Biologicals is a mouse monoclonal antibody to SSH1. This antibody reacts with human. The SSH1 Antibody (1D2) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SSH1
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR