Anti-SSEA-3 Rat Monoclonal Antibody (DyLight 680) [clone: MC-631]

NB100-1832FR
103368-710EA 466.15 USD
103368-710
Anti-SSEA-3 Rat Monoclonal Antibody (DyLight 680) [clone: MC-631]
The SSEA-3 Antibody (MC-631) [DyLight 680] from Novus Biologicals is a rat monoclonal antibody to SSEA-3. This antibody reacts with human, mouse. The SSEA-3 Antibody (MC-631) [DyLight 680] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: SSEA-3
Clonality: Monoclonal
Clone: MC-631
Conjugation: DyLight 680
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR