Anti-SSC5D Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-93831
103283-478EA 734.58 USD
103283-478
Anti-SSC5D Rabbit Polyclonal Antibody
Antibodies
The SSC5D Antibody from Novus Biologicals is a rabbit polyclonal antibody to SSC5D. This antibody reacts with human. The SSC5D Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SSC5D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR