Anti-SSBP2 Mouse Monoclonal Antibody [clone: 2E7]

Supplier: Novus Biologicals
H00023635-M10
103341-650EA 798.46 USD
103341-650
Anti-SSBP2 Mouse Monoclonal Antibody [clone: 2E7]
Antibodies
The SSBP2 Antibody (2E7) from Novus Biologicals is a mouse monoclonal antibody to SSBP2. This antibody reacts with human. The SSBP2 Antibody (2E7) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SSBP2
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR