Anti-SSB Mouse Monoclonal Antibody [clone: 1D6-H5]

Supplier: Novus Biologicals
H00006741-M01
103310-788EA 798.46 USD
103310-788
Anti-SSB Mouse Monoclonal Antibody [clone: 1D6-H5]
Antibodies
The SSB Antibody (1D6-H5) from Novus Biologicals is a mouse monoclonal antibody to SSB. This antibody reacts with human. The SSB Antibody (1D6-H5) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SSB
Clonality: Monoclonal
Clone: 1D6-H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR