Anti-SS18L2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13386
103285-758EA 734.58 USD
103285-758
Anti-SS18L2 Rabbit Polyclonal Antibody
Antibodies
The SS18L2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SS18L2. This antibody reacts with human. The SS18L2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SS18L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR