Anti-SRY Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00006736-B01P
103310-770EA 798.46 USD
103310-770
Anti-SRY Mouse Polyclonal Antibody
Antibodies
The SRY Antibody from Novus Biologicals is a mouse polyclonal antibody to SRY. This antibody reacts with human. The SRY Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SRY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR