Anti-SRRM1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13381
103285-748EA 734.58 USD
103285-748
Anti-SRRM1 Rabbit Polyclonal Antibody
Antibodies
The SRRM1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SRRM1. This antibody reacts with human. The SRRM1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SRRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR