Anti-SRPK1 Mouse Monoclonal Antibody [clone: 1D11]

Supplier: Novus Biologicals
H00006732-M11
103310-760EA 399 USD
103310-760
Anti-SRPK1 Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
The SRPK1 Antibody (1D11) from Novus Biologicals is a mouse monoclonal antibody to SRPK1. This antibody reacts with human. The SRPK1 Antibody (1D11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SRPK1
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR